ContactUsLinkComponent
ReadiLink™ Rapid iFluor® 750 Antibody Labelling Kit
ReadiLink™ Rapid iFluor® 750 Antibody Labelling Kit
Catalog # 77462-916
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® 750 Antibody Labelling Kit
Catalog # 77462-916
Supplier:  AAT BIOQUEST INC
Supplier Number:  5718
CAS Number:  
Specifications
  • Size:
    3×1 mg
  • Cat. No.:
    77462-916
Specifications
About this item
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with our superior iFluor® dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.