ContactUsLinkComponent
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Catalog # 77461-454
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Catalog # 77461-454
Supplier:  AAT BIOQUEST INC
Supplier Number:  1348
CAS Number:  
Specifications
  • Size:
    2
  • Cat. No.:
    77461-454
  • Includes:
    2 labellings
Specifications
About this item
Microscale optimized for labelling 25 µg antibody per reaction. APC-iFluor® 750 tandem is an excellent replacement for APC-Cy7 and APC-Alexa Fluor® 750 tandem since they have almost identical spectra.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.