ContactUsLinkComponent
Amplite® Fluorimetric Lithium Ion Quantification Kit
Amplite® Fluorimetric Lithium Ion Quantification Kit
Catalog # 77461-876
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Amplite® Fluorimetric Lithium Ion Quantification Kit
Catalog # 77461-876
Supplier:  AAT BIOQUEST INC
Supplier Number:  21351
CAS Number:  
Specifications
  • Size:
    100 Tests
  • Cat. No.:
    77461-876
  • No. of tests:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. Amplite® fluorimetric lithium ion quantification kit uses our new robust lithium-ion indicator dye, Lithiumighty™ 520, which exhibits great fluorescence intensity enhancement upon binding to lithium ions.
  • Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
  • Requires very small amount of sample
  • 12 month shelf life
Microplate-based assay kit is particularly suitable for the determination of lithium ion concentration in a microvolume format.
Delivery: Shipped at RT.
Caution: Research use only.