ContactUsLinkComponent
JOE Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Catalog # 77462-998
Supplier:  AAT Bioquest
undefined
JOE Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Catalog # 77462-998
Supplier:  AAT Bioquest
Supplier Number:  67012
Specifications
  • Frame color:
    White
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Semi-Skirted
  • Well Color:
    White
  • Description:
    JOE Dye qPCR calibration plate
  • Cat. No.:
    77462-998
Specifications
About this item
The JOE dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than three times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimize light exposure.