ContactUsLinkComponent
iFluor® 350-Wheat Germ Agglutinin (WGA) Conjugate
iFluor® 350-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-026
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 350-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-026
Supplier:  AAT BIOQUEST INC
Supplier Number:  25525
CAS Number:  
Specifications
  • Description:
    iFluor® 350-Wheat Germ Agglutinin (WGA) conjugate
  • Size:
    1 mg
  • Cat. No.:
    77462-026
Specifications
About this item
iFluor® 350 WGA conjugate, like Alexa Fluor® 350 WGA conjugate, retains its ability to bind to sialic acid and N-acetylglucosaminyl residues, enhancing its utility in fluorescence imaging and analysis for various scientific investigations.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.