HIS Lite™ Cy3 Tris NTA-Ni Complex
Catalog # 77461-402
Supplier: AAT BIOQUEST INC
CAS Number:
Specifications
- Description:HIS Lite™ Cy3 Tris NTA-Ni complex
- Size:100 μg
- Cat. No.:77461-402
Specifications
About this item
Cy3-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labeled proteins in cells, solution and solid surfaces.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.