Gelite™ 50 bp DNA Ladder
77461-670
:
- Description:Gelite™ 50 bp DNA ladder
- Size:0.5 ml
- Cat. No.:77461-670
The Gelite™ 50 bp DNA Ladder is ideal for sizing and approximating the mass of double-stranded DNA within the 50 to 1500 bp range on agarose or polyacrylamide gels.
- Stable for 12 months
Quality tested in-house.
: For research use only.