Human Recombinant IRF4 (from E. coli)
Vial, 50 µg
Catalog # 77540-030
Supplier: Creative Biomart
Supplier #: IRF4-106H
Human Recombinant IRF4 (from <i>E. coli</i>)
Catalog # 77540-030
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:50 µg
- Storage Conditions:4 °C
- Molecular Weight:The protein has a calculated MW of 55 kDa
- Protease-free:No
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.5 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin Level:Low
- Animal-Free:No
- UniProtKB:Q15306
- Purity:>90% as determined by SDS-PAGE
- Carrier-Free:No
- Grade:Ultrapure grade
- Endotoxin Content:<1.0 EU/µg by the LAL method
- Source:E. coli
- Nuclease-free:No
- Cat. No.:77540-030
- Shipping Temperature:20
- Protein/Peptide Type:Recombinant
- Protein Synonyms:interferon regulatory factor 4
- Protein/Peptide Name:IRF4
- Conjugation:Unconjugated
- Endotoxin-free:No
- Species:Human



