Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ligase (from E. coli)

Vial, 1000 U
Catalog # 77539-096
Supplier:   Creative Biomart
 :  CMC12
Ligase (from <i>E. coli</i>)
Catalog # 77539-096
Supplier:  Creative Biomart
CAS Number:  
Ligase (from <i>E. coli</i>)
Ligase (from E. coli)

Vial, 1000 U

CAS Number:  
Price
$530.89
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    1000 U
  • Storage Conditions:
    4 °C
  • Protease-free:
    No
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • Tested Applications:
    Cloning of restriction fragments, Joining linkers and adapters to blunt-ended DNA
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Concentration:
    400000 units/ml
  • Source:
    E. coli
  • Nuclease-free:
    No
  • Cat. No.:
    77539-096
  • Shipping Temperature:
    20
  • Protein Synonyms:
    Ligase
  • Protein/Peptide Name:
    Ligase
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No

Specifications