Human Recombinant BMP2 (from E. coli)
Vial, 200 µg
Catalog # 77538-752
Supplier: Creative Biomart
Supplier #: BMP2-26315TH
Human Recombinant BMP2 (from <i>E. coli</i>)
Catalog # 77538-752
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:200 µg
- Storage Conditions:4 °C
- Protease-free:No
- Endotoxin Level:Low
- Animal-Free:No
- UniProtKB:P12643
- Purity:>95% by SDS-PAGE
- Carrier-Free:No
- Grade:Ultrapure grade
- Source:E. coli
- Nuclease-free:No
- Cat. No.:77538-752
- Shipping Temperature:20
- Protein/Peptide Type:Recombinant
- Protein Synonyms:bone morphogenetic protein 2
- Protein/Peptide Name:BMP2
- Conjugation:Unconjugated
- Endotoxin-free:No
- Species:Human



