Human Recombinant LTF (from Rice flour)
77540-174
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Rice flour
- Species:Human
- Size:100 mg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:lactotransferrin
- UniProtKB:P02788
- Protein/Peptide Name:LTF
- Purity:>90% as determined by: Analysis by RP-HPLC and analysis by SDS-PAGE
- Molecular Weight:77 - 80 kDa
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77540-174