Human Recombinant EIF3M (from E. coli)
Vial, 100 µg
Catalog # 77539-358
Supplier: Creative Biomart
Supplier #: EIF3M-392H
Human Recombinant EIF3M (from <i>E. coli</i>)
Catalog # 77539-358
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:100 µg
- Storage Conditions:4 °C
- Protease-free:No
- Endotoxin Level:Low
- Animal-Free:No
- UniProtKB:Q7L2H7
- Purity:>90% (SDS-PAGE)
- Carrier-Free:No
- Grade:Ultrapure grade
- Sequence:SVPAFIDIS EEDQAAELRA YLKSKGAEIS EENSEGGLHV DLAQIIEACD VCLKEDDKDV ESVMNSVVSL LLILEPDKQE ALIESLCEKL VKFREGERPS LRLQLLSNLF HGMDKNTPVR YTVYCSLIKV AASCGAIQYI PTELDQVRKW ISDWNLTTEK KHTLLRLLYE ALVDCKKSDA ASKVMVELLG SYTEDNASQA RVDAHRCIVR ALKDPNAFLF DHLLTLKPVK FLEGELIHDL LTIFVSAKLA SYVKFYQNNK DFIDSLGLLH EQNMAKMRLL TFMGMAVENK EISFDTMQQE LQIGADDVEA FVIDAVRTKM VYCKIDQTQR KVVVSHSTHR TFGKQQWQQL YDTLNAWKQN LNKVKNSLLS LSDT
- Source:E. coli
- Nuclease-free:No
- Cat. No.:77539-358
- Shipping Temperature:20
- Protein/Peptide Type:Recombinant
- Protein Synonyms:eukaryotic translation initiation factor 3, subunit M
- Protein/Peptide Name:EIF3M
- Conjugation:Unconjugated
- Endotoxin-free:No
- Species:Human
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



