Human Recombinant ACVR2A
Vial, 100 µg
Catalog # 77538-552
Supplier: Creative Biomart
Supplier #: ACVR2A-P015H-100UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:100 µg
- Storage Conditions:4 °C
- Protease-free:No
- Endotoxin Level:Low
- Animal-Free:No
- UniProtKB:P27037
- Purity:>95%
- Carrier-Free:No
- Grade:Ultrapure grade
- Endotoxin Content:<0.1 EU/μg of the protein
- Nuclease-free:No
- Cat. No.:77538-552
- Shipping Temperature:Blue ice
- Protein/Peptide Type:Recombinant
- Protein Synonyms:activin receptor IIA
- Protein/Peptide Name:ACVR2A
- Conjugation:Unconjugated
- Endotoxin-free:No
- Species:Human



