ContactUsLinkComponent
ProLite™ 5 to 245 kD Protein Ladder
ProLite™ 5 to 245 kD Protein Ladder
Catalog # 77461-702
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
ProLite™ 5 to 245 kD Protein Ladder
Catalog # 77461-702
Supplier:  AAT BIOQUEST INC
Supplier Number:  18015
CAS Number:  
Specifications
  • Description:
    ProLite™ 5-245 kD protein ladder
  • Size:
    100 µl
  • Environmentally Preferable:
  • Cat. No.:
    77461-702
Specifications
About this item
The ProLite™ 5 to 245 kD protein ladder is a high-quality three-colour protein standard consisting of 13 prestained proteins.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.