ContactUsLinkComponent
NED Dye qPCR Calibration Solution *10000X*
NED Dye qPCR Calibration Solution *10000X*
Catalog # 77463-008
Supplier:  AAT Bioquest
CAS Number:  
undefined
NED Dye qPCR Calibration Solution *10000X*
Catalog # 77463-008
Supplier:  AAT Bioquest
Supplier Number:  67031
CAS Number:  
Specifications
  • Description:
    NED Dye qPCR calibration solution *10000X*
  • Size:
    100 µl
  • Cat. No.:
    77463-008
Specifications
About this item
NED dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.