ContactUsLinkComponent
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-010
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-010
Supplier:  AAT BIOQUEST INC
Supplier Number:  25450
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    mFluor™ Violet 450-Wheat germ agglutinin (WGA) conjugate
  • Size:
    1 mg
  • Cat. No.:
    77462-010
Specifications
About this item
mFluor™ Violet 450 labeled WGA is well-excited by the violet laser, emitting a bright blue fluorescence at 445 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.