Orders LinkContactUsLinkComponent
VEE GEE Volumetric Pipettes
VEE GEE Volumetric Pipettes
VEE GEE Volumetric Pipettes
VEE GEE Volumetric Pipettes
VEE GEE Scientific's volumetric pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of your liquid handling needs.
  • Made out of high-quality borosilicate glass
  • Wide range of sizes to choose from
  • Reusable
  • Color-coded with permanent enamel markings
  • Calibrated to deliver
  • Meets ASTM E-969 Class A specifications and USP standards
Our volumetric pipettes are graduated to the tip and made out of high-quality borosilicate glass. These pipettes also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
From micro to macro, our line of glass volumetric pipettes cover a wide range of volumes, making VEE GEE the perfect one-stop shop for all of your pipetting needs.
Order your volumetric pipettes with us online today and receive free shipping and fast delivery.
Product Family Options
  • Capacity
    Length
    Color code
    Tolerance
  • 2 ml
    13.9"
    Orange
    0.006 ml
    Pack of 12
    Pack of 12 - $49.34
    $49.34
    Case of 864
    Case of 864 - $3,552.48
    $3,552.48
    Pack of 12
       $49.34
    Case of 864
       $3,552.48
     : 2040A-2
  • 3 ml
    14.1"
    Black
    0.01 ml
    Pack of 12
    Pack of 12 - $57.77
    $57.77
    Case of 864
    Case of 864 - $4,159.44
    $4,159.44
    Pack of 12
       $57.77
    Case of 864
       $4,159.44
     : 2040A-3
  • 5 ml
    14.7"
    White
    0.01 ml
    Pack of 12
    Pack of 12 - $65.60
    $65.60
    Case of 864
    Case of 864 - $4,723.20
    $4,723.20
    Pack of 12
       $65.60
    Case of 864
       $4,723.20
     : 2040A-5
  • 6 ml
    15.3"
    Orange
    0.01 ml
    Pack of 12
    Pack of 12 - $111.45
    $111.45
    Case of 864
    Case of 864 - $8,024.40
    $8,024.40
    Pack of 12
       $111.45
    Case of 864
       $8,024.40
     : 2040A-6
  • 8 ml
    16.9"
    Blue
    0.02 ml
    Pack of 12
    Pack of 12 - $117.00
    $117.00
    Case of 864
    Case of 864 - $8,424.00
    $8,424.00
    Pack of 12
       $117.00
    Case of 864
       $8,424.00
     : 2040A-8
  • 10 ml
    17.3"
    Red
    0.02 ml
    Pack of 12
    Pack of 12 - $117.00
    $117.00
    Case of 864
    Case of 864 - $8,424.00
    $8,424.00
    Pack of 12
       $117.00
    Case of 864
       $8,424.00
     : 2040A-10
  • 13 ml
    18.1"
    White
    0.03 ml
    Pack of 6
    Pack of 6 - $62.49
    $62.49
    Case of 216
    Case of 216 - $2,249.64
    $2,249.64
    Pack of 6
       $62.49
    Case of 216
       $2,249.64
     : 2040A-13
  • 15 ml
    18.1"
    Green
    0.03 ml
    Pack of 6
    Pack of 6 - $58.52
    $58.52
    Case of 216
    Case of 216 - $2,106.72
    $2,106.72
    Pack of 6
       $58.52
    Case of 216
       $2,106.72
     : 2040A-15
  • 25 ml
    18.8"
    Blue
    0.03 ml
    Pack of 6
    Pack of 6 - $62.79
    $62.79
    Case of 216
    Case of 216 - $2,260.44
    $2,260.44
    Pack of 6
       $62.79
    Case of 216
       $2,260.44
     : 2040A-25