Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Chicken Native LYZ (from Egg)

Chicken Native LYZ (from Egg)

Vial, 1 g
Catalog # 77540-188
Supplier:   Creative Biomart
Supplier #:  LYZ-65E
Chicken Native LYZ (from Egg)
Catalog # 77540-188
Supplier:  Creative Biomart
CAS Number:  
Chicken Native LYZ (from Egg)
Chicken Native LYZ (from Egg)

Vial, 1 g

CAS Number:  
Price
$52.24
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    1 g
  • Storage Conditions:
    4 °C
  • Protease-free:
    No
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • UniProtKB:
    P00698
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Source:
    Egg
  • Nuclease-free:
    No
  • Cat. No.:
    77540-188
  • Shipping Temperature:
    20
  • Protein/Peptide Type:
    Native
  • Protein Synonyms:
    lysozyme
  • Protein/Peptide Name:
    LYZ
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No
  • Species:
    Chicken

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.