Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human Recombinant CASP4 (from E. coli)

Supplier:   Creative Biomart
Human Recombinant CASP4 (from <i>E. coli</i>)
Supplier:  Creative Biomart
Certificates
Human Recombinant CASP4 (from <i>E. coli</i>)

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77538-834
  • 77538-832
  • Protein/Peptide Name:
  • CASP4
  • CASP4
  • Protein Synonyms:
  • caspase 4, apoptosis-related cysteine peptidase
  • caspase 4, apoptosis-related cysteine peptidase
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Protease-free:
  • No
  • No
  • Animal-Free:
  • No
  • No
  • Nuclease-free:
  • No
  • No
  • Carrier-Free:
  • No
  • No
  • Endotoxin-free:
  • No
  • No
  • Endotoxin Level:
  • Low
  • Low
  • UniProtKB:
  • P49662
  • P49662
  • Purity:
  • ≥95% (SDS-PAGE)
  • ≥95% (SDS-PAGE)
  • Grade:
  • Ultrapure grade
  • Ultrapure grade
  • Reconstitution Instructions:
  • Reconstitute with PBS containing 15% glycerol to 1 U/µl
  • Reconstitute with PBS containing 15% glycerol to 1 U/µl
  • Storage Conditions:
  • 4 °C
  • 4 °C
  • Shipping Temperature:
  • 20
  • 20

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    10 U
    Log in for availability
    Log in for availability
    Each
    Each - $211.08
    $211.08
    Each
       $211.08
     

    Supplier Number: CASP4-599H-10U
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77538-834
    Protein/Peptide Name
    CASP4
    Protein Synonyms
    caspase 4, apoptosis-related cysteine peptidase
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Protease-free
    No
    Animal-Free
    No
    Nuclease-free
    No
    Carrier-Free
    No
    Endotoxin-free
    No
    Endotoxin Level
    Low
    UniProtKB
    P49662
    Purity
    ≥95% (SDS-PAGE)
    Grade
    Ultrapure grade
    Reconstitution Instructions
    Reconstitute with PBS containing 15% glycerol to 1 U/µl
    Storage Conditions
    4 °C
    Shipping Temperature
    20
  • Vial
    100 U
    Log in for availability
    Log in for availability
    Each
    Each - $637.50
    $637.50
    Each
       $637.50
     

    Supplier Number: CASP4-599H-100U
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77538-832
    Protein/Peptide Name
    CASP4
    Protein Synonyms
    caspase 4, apoptosis-related cysteine peptidase
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Protease-free
    No
    Animal-Free
    No
    Nuclease-free
    No
    Carrier-Free
    No
    Endotoxin-free
    No
    Endotoxin Level
    Low
    UniProtKB
    P49662
    Purity
    ≥95% (SDS-PAGE)
    Grade
    Ultrapure grade
    Reconstitution Instructions
    Reconstitute with PBS containing 15% glycerol to 1 U/µl
    Storage Conditions
    4 °C
    Shipping Temperature
    20