Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Native ACTB (from Phlatelets)

Human Native ACTB (from Phlatelets)

Vial, 25 µg
Catalog # 77538-546
Supplier:   Creative Biomart
Supplier #:  ACTB-325H
Human Native ACTB (from Phlatelets)
Catalog # 77538-546
Supplier:  Creative Biomart
CAS Number:  
Human Native ACTB (from Phlatelets)
Human Native ACTB (from Phlatelets)

Vial, 25 µg

CAS Number:  
Price
$340.07
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    25 µg
  • Storage Conditions:
    4 °C
  • Molecular Weight:
    Non-muscle actin has an approximate molecular weight of 43 kDa
  • Protease-free:
    No
  • Reconstitution Instructions:
    The protein should be reconstituted to 10 mg/ml with 100 µl of distilled water, it will then be in the following buffer: 5 mM Tris-HCl pH 8.0, 0.2 mM CaCl₂, 0.2 mM ATP, 5% (w/v) sucrose, and 1% (w/v) dextran.
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • UniProtKB:
    P60709
  • Purity:
    Protein purity is determined by scanning densitometry of Coomassie blue stained protein on a 12% polyacrylamide gel. Non-muscle actin was found to be >99% pure.
  • Tested Applications:
    Identification and characterization of non-muscle actin binding proteins,<i>in vitro</i> actin polymerization studies,antibody standard for western blot analysis
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Source:
    Phlatelets
  • Nuclease-free:
    No
  • Cat. No.:
    77538-546
  • Shipping Temperature:
    Blue ice
  • Biological Activity:
    The biological activity of non-muscle actin can be determined by its ability to efficiently polymerize into filaments in vitro and separate from unpolymerized components in a spin down assay. Stringent quality control ensures that >85% of the non-muscle actin can polymerized in this assay.
  • Protein/Peptide Type:
    Native
  • Protein Synonyms:
    actin, beta
  • Protein/Peptide Name:
    ACTB
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.