Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Recombinant Human GPC4 (from <i>E. coli</i>)

Recombinant Human GPC4 (from E. coli)

Supplier:   Creative Biomart
Recombinant Human GPC4 (from <i>E. coli</i>)
Supplier:  Creative Biomart
Certificates
Recombinant Human GPC4 (from <i>E. coli</i>)

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77539-630
  • 77539-628
  • Protein/Peptide Name:
  • GPC4
  • GPC4
  • Protein Synonyms:
  • glypican 4
  • glypican 4
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Sequence:
  • MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS
  • MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS
  • Protease-free:
  • No
  • No
  • Animal-Free:
  • No
  • No
  • Nuclease-free:
  • No
  • No
  • Carrier-Free:
  • No
  • No
  • Endotoxin-free:
  • No
  • No
  • Endotoxin Level:
  • Low
  • Low
  • UniProtKB:
  • O75487
  • O75487
  • Purity:
  • >85% by SDS-PAGE
  • >85% by SDS-PAGE
  • Grade:
  • Ultrapure grade
  • Ultrapure grade
  • Concentration:
  • 0.25 mg/ml (determined by Bradford assay)
  • 0.25 mg/ml (determined by Bradford assay)
  • Molecular Weight:
  • 18 kDa (165aa) confirmed by MALDI-TOF(Molecular size on SDS-PAGE will appear higher)
  • 18 kDa (165aa) confirmed by MALDI-TOF(Molecular size on SDS-PAGE will appear higher)
  • Storage Conditions:
  • 4 °C
  • 4 °C
  • Shipping Temperature:
  • 20
  • 20

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    5 µg
    Log in for availability
    Log in for availability
    Each
    Each - $104.47
    $104.47
    Each
       $104.47
    Minimum order quantity is 1

    Supplier Number: GPC4-519H-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77539-630
    Protein/Peptide Name
    GPC4
    Protein Synonyms
    glypican 4
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Sequence
    MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS
    Protease-free
    No
    Animal-Free
    No
    Nuclease-free
    No
    Carrier-Free
    No
    Endotoxin-free
    No
    Endotoxin Level
    Low
    UniProtKB
    O75487
    Purity
    >85% by SDS-PAGE
    Grade
    Ultrapure grade
    Concentration
    0.25 mg/ml (determined by Bradford assay)
    Molecular Weight
    18 kDa (165aa) confirmed by MALDI-TOF(Molecular size on SDS-PAGE will appear higher)
    Storage Conditions
    4 °C
    Shipping Temperature
    20
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    Each
    Each - $2,129.97
    $2,129.97
    Each
       $2,129.97
    Minimum order quantity is 1

    Supplier Number: GPC4-519H-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77539-628
    Protein/Peptide Name
    GPC4
    Protein Synonyms
    glypican 4
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Sequence
    MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS
    Protease-free
    No
    Animal-Free
    No
    Nuclease-free
    No
    Carrier-Free
    No
    Endotoxin-free
    No
    Endotoxin Level
    Low
    UniProtKB
    O75487
    Purity
    >85% by SDS-PAGE
    Grade
    Ultrapure grade
    Concentration
    0.25 mg/ml (determined by Bradford assay)
    Molecular Weight
    18 kDa (165aa) confirmed by MALDI-TOF(Molecular size on SDS-PAGE will appear higher)
    Storage Conditions
    4 °C
    Shipping Temperature
    20
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.