Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Recombinant Human DFFA (from Human)

Vial, 10 µg
Catalog # 77539-286
Supplier:   Creative Biomart
 :  DFFA-4316H
Recombinant Human DFFA (from Human)
Catalog # 77539-286
Supplier:  Creative Biomart
CAS Number:  
Recombinant Human DFFA (from Human)

Vial, 10 µg

CAS Number:  
Price
$233.46
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    10 µg
  • Storage Conditions:
    4 °C
  • Protease-free:
    No
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • UniProtKB:
    O00273
  • Purity:
    >90.0% as determined by SDS-PAGE
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Source:
    Human cells
  • Nuclease-free:
    No
  • Cat. No.:
    77539-286
  • Shipping Temperature:
    20
  • Protein/Peptide Type:
    Recombinant
  • Protein Synonyms:
    DNA fragmentation factor, 45kDa, alpha polypeptide
  • Protein/Peptide Name:
    DFFA
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No
  • Species:
    Human

Specifications