Native Human C3d (from Plasma)
Supplier: Creative Biomart
About this item
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77539-116
- 77539-118
- 77539-114
- Protein/Peptide Name:
- C3d
- C3d
- C3d
- Protein Synonyms:
- Complement C3d
- Complement C3d
- Complement C3d
- Protein/Peptide Type:
- Native
- Native
- Native
- Species:
- Human
- Human
- Human
- Source:
- Plasma
- Plasma
- Plasma
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Protease-free:
- No
- No
- No
- Animal-Free:
- No
- No
- No
- Nuclease-free:
- No
- No
- No
- Carrier-Free:
- No
- No
- No
- Endotoxin-free:
- No
- No
- No
- Endotoxin Level:
- Low
- Low
- Low
- Purity:
- >95% by SDS-PAGE
- >95% by SDS-PAGE
- >95% by SDS-PAGE
- Grade:
- Ultrapure grade
- Ultrapure grade
- Ultrapure grade
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- 1.0 mg/ml
- Molecular Weight:
- 33.8 kDa (single chain)
- 33.8 kDa (single chain)
- 33.8 kDa (single chain)
- Storage Conditions:
- 4 °C
- 4 °C
- 4 °C
- Shipping Temperature:
- 20
- 20
- 20
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: COMPLEMENTC3D-48HB-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-116Protein/Peptide NameC3dProtein SynonymsComplement C3dProtein/Peptide TypeNativeSpeciesHumanSourcePlasmaConjugationUnconjugatedProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowPurity>95% by SDS-PAGEGradeUltrapure gradeConcentration1.0 mg/mlMolecular Weight33.8 kDa (single chain)Storage Conditions4 °CShipping Temperature20 - Supplier Number: COMPLEMENTC3D-48HB-25UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-118Protein/Peptide NameC3dProtein SynonymsComplement C3dProtein/Peptide TypeNativeSpeciesHumanSourcePlasmaConjugationUnconjugatedProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowPurity>95% by SDS-PAGEGradeUltrapure gradeConcentration1.0 mg/mlMolecular Weight33.8 kDa (single chain)Storage Conditions4 °CShipping Temperature20 - Supplier Number: COMPLEMENTC3D-48HB-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-114Protein/Peptide NameC3dProtein SynonymsComplement C3dProtein/Peptide TypeNativeSpeciesHumanSourcePlasmaConjugationUnconjugatedProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowPurity>95% by SDS-PAGEGradeUltrapure gradeConcentration1.0 mg/mlMolecular Weight33.8 kDa (single chain)Storage Conditions4 °CShipping Temperature20



