


Toluene-[D₈] 100.0 atom % D for NMR spectroscopy
Catalog # TS217440050
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass ampoule
- Size:5 ml
- Cat. No.:TS217440050
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.01 %
- D-Enrichment:≥99.95 %
Specification Test Results
About this item
:
:100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638