Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Toxic
Health
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Catalog # TS217310100
New Product
CAS Number:  
Toxic
Health
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Catalog # TS217310100
Supplier Number:  217310100
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Size:
    10 ml
  • Cat. No.:
    TS217310100

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

About this item

 :
 :120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827