Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Digoxigenin C9 acid

5 mg
Catalog # 77462-196
Supplier:   AAT Bioquest
 :  3510
Digoxigenin C9 acid
Catalog # 77462-196
Supplier:  AAT Bioquest
CAS Number:  
Digoxigenin C9 acid
Digoxigenin C9 acid

5 mg

CAS Number:  
Price
$329.63
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:561.72 g/mol
Specifications

Specifications

  • Size:
    5 mg

Specifications