Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cy7 biotin conjugate

Cy7 biotin conjugate

5 mg
Catalog # 77462-180
Supplier:   AAT Bioquest
Supplier #:  3106
Cy7 biotin conjugate
Catalog # 77462-180
Supplier:  AAT Bioquest
CAS Number:  
Cy7 biotin conjugate
Cy7 biotin conjugate

5 mg

CAS Number:  
Price
$218.19
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cy7 biotin conjugate demonstrates excellent avidin binding and strong NIR fluorescence.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:1291.49 g/mol
Specifications

Specifications

  • Size:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.