Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AzoDye-2 CPG
AzoDye-2 CPG (500 A)
AzoDye-2 CPG (500 A)
AzoDye-2 CPG (500 A)
Catalog # 77461-968
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
AzoDye-2 CPG (500 A)
Catalog # 77461-968
Supplier:
AAT BIOQUEST INC
Supplier Number:
2428
CAS Number:
Specifications
Size:
100 mg
Environmentally Preferable:
Cat. No.:
77461-968
Specifications
About this item
Product Details
:
Product Details