Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ac-DEVD-aminoluciferin

1 mg
Catalog # 77461-436
Supplier:   AAT Bioquest
 :  13210
Ac-DEVD-aminoluciferin
Catalog # 77461-436
Supplier:  AAT Bioquest
CAS Number:  
Ac-DEVD-aminoluciferin
Ac-DEVD-aminoluciferin

1 mg

CAS Number:  
Price
$103.21
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ac-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.

  • Stable for 24 months

Quality tested in-house

 : For Research Use Only

  • Formula:C₃₁H₃₇N₇O₁₃S₂
  • Molecular Weight:779.79 g/mol
Specifications

Specifications

  • Size:
    1 mg

Specifications