UltraTemp® ESD Polyamide Circuit Board Labels for Printers
Supplier: Brady Worldwide
New Product
Certificates
About this item
Ideal for PCB and electronic component manufacturing, these labels are static dissipative and survive extreme processes like wave soldering and multiple harsh washes.
- Engineered to withstand high temperatures
- Low profile, glossy white polyimide material (B-767)
- Static dissipative adhesive and liner
- Labels do not require reflow or preheatings to ensure printing readability
- Service temperature of −70° to +100 °C (−94 to +212 °F)
Enhanced topcoat is ideal for boards that go through multiple harsh-condition washes. A ribbon is required to print this material. Surface resistivity meets ANSI/ESD S541-2008 recommended range for dissipative ESD packaging materials.
Certifications: UL approved, meets requirements of MIL-STD-202G, method 215K.