Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ca²⁺/Calmodulin Solution II, 10X

Ca²⁺/Calmodulin Solution II, 10X, 500 µl, –20 °C
Catalog # 77554-936
Supplier:   Sino Biological
 :  C02-39B-500
Ca²⁺/Calmodulin Solution II, 10X
Catalog # 77554-936
Supplier:  Sino Biological
CAS Number:  
Ca²⁺/Calmodulin Solution II, 10X

Ca²⁺/Calmodulin Solution II, 10X, 500 µl, –20 °C

CAS Number:  
Price
$17.40
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

10X stock solution used for assaying activity of calcium-calmodulin-activated protein kinase enzymes.

Specifications

Specifications

  • Description:
    Ca²⁺/Calmodulin Solution II, 10X
  • Size:
    500 µl
  • Storage temperature:
    –20 °C
  • Application:
    Kinase assay
  • Storage Conditions:
    For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Cat. No.:
    77554-936
  • Formulation:
    Frozen solution of buffer containing 50 ng/µl Calmodulin, 4 mM CaCl₂, 10 mM Tris-HCl, pH 7.2

Specifications