Order Entry
Puerto Rico
ContactUsLinkComponent
PermaShield™ Matte Polyester Label for BradyJet™ J2000 Printers
PermaShield™ Matte Polyester Label for BradyJet™ J2000 Printers
Catalog # 77574-948
Supplier:  Brady Worldwide
New Product
undefined
PermaShield™ Matte Polyester Label for BradyJet™ J2000 Printers
Catalog # 77574-948
Supplier:  Brady Worldwide
Supplier Number:  175617
New Product

Specifications

  • For Use With:
    J2000
  • Color:
    White
  • Dimensions:
    1H×2"W (1.5H×2.5"W overlaminate)
  • Material:
    Inkjet receptive polyester
  • Thickness:
    0.0047"
  • Cat. No.:
    77574-948
  • Packaging:
    750 labels/roll

Specifications

About this item

Upgrade the durability and longevity of your identification projects, even in the toughest environments, with PermaShield™ polyester labels designed for BradyJet printers.

  • Simply print label, adhere to surface and apply the matching overlaminate
  • Commonly used to print product labeling, logos, right to know labels and barcode applications
  • Works best on flat, smooth surfaces, indoor use
  • An ink cartridge is required to print this material; use part number J20-CMY
  • Abrasion-resistant, chemical-resistant, fade-resistant, oil-resistant, solvent-resistant

The durable polyester label material (B-2475) plus polyester overlaminate (B-996) protects against numerous harsh solvents and provides a more glossy appearance.

Delivery: PermaShield by Brady is a 2-in-1 package that includes a printable label plus matching clear polyester overlaminate.