ContactUsLinkComponent
Fluorescein-12-dGTP *1 mM in Tris Buffer (pH 7.5)*
Fluorescein-12-dGTP *1 mM in Tris Buffer (pH 7.5)*
Catalog # 77461-522
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Fluorescein-12-dGTP *1 mM in Tris Buffer (pH 7.5)*
Catalog # 77461-522
Supplier:  AAT BIOQUEST INC
Supplier Number:  17037
CAS Number:  
Specifications
  • Size:
    25 nmoles
  • Cat. No.:
    77461-522
Specifications
About this item
Fluorescein-12-dGTP might be incorporated into a DNA sequence as its natural counterpart dGTP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labeling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and guanosine has been optimized for the optimal substrate incorporation and labeling efficiency.
Caution: For research use only.