ContactUsLinkComponent
DEAC-dUTP *1 mM in TE Buffer (pH 7.5)*
DEAC-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-516
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
DEAC-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-516
Supplier:  AAT BIOQUEST INC
Supplier Number:  17034
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    100 nmoles
  • Cat. No.:
    77461-516
Specifications
About this item
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
  • Chemically identical to Diethylaminocoumarin-5-dUTP (Perkin Elmer’s NEL455001EA)
This Coumarin-dUTP conjugate can be used as a blue fluorescence color with Spectrum Aqua™ filter set (Spectrum Aqua™ is the trademark of Vysis).
Caution: For research use only.