ContactUsLinkComponent
ReadiUse™ Yeast Cell Lysis Buffer, 5X
ReadiUse™ Yeast Cell Lysis Buffer, 5X
Catalog # 77461-950
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
ReadiUse™ Yeast Cell Lysis Buffer, 5X
Catalog # 77461-950
Supplier:  AAT BIOQUEST INC
Supplier Number:  24102
CAS Number:  
Specifications
  • Description:
    ReadiUse™ Yeast cell lysis buffer 5X
  • Cat. No.:
    77461-950
  • Volume:
    10 ml
Specifications
About this item
ReadiUse™ Yeast Cell Lysis Buffer is a biochemical reagent designed for efficient yeast cell lysis and protein solubilization in laboratory applications.
  • Designed for ease of use
  • Saves time
  • Versatile in its efficacy across various organisms, including Bacillus subtilis, Gram-positive, and Gram-negative bacteria
With a mild detergent formulation, ReadiUse™ Yeast cell lysis buffer minimizes the reliance on harsh chemical treatments, extreme pH adjustments, or temperature variations, contributing to the preservation of extracted protein integrity.
Caution: For research use only.