ContactUsLinkComponent
PBS Tablet (Phosphate Buffered Saline Tablet)
PBS Tablet (Phosphate Buffered Saline Tablet)
Catalog # 77461-346
Supplier:  AAT Bioquest
CAS Number:  
undefined
PBS Tablet (Phosphate Buffered Saline Tablet)
Catalog # 77461-346
Supplier:  AAT Bioquest
Supplier Number:  10010
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    10 tablets
  • Description:
    Phosphate buffered saline tablet
  • Cat. No.:
    77461-346
Specifications
About this item
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
  • Designed for ease of use
  • Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Caution: For research use only.