HPLC LISA Waste Bundle, Type 20
Supplier: SCAT AMERICAS, INC.
Jump to:
About this item
HPLC LISA Waste bundle, type 20 with and without funnel.
Certifications: UN approved.
Delivery information: Kit includes 1×Safety waste cap LISA, S60/61, 1×20 L canister, S60/61, PE-HD, 1×exhaust filter M, with splash guard and change indicator.
2 Options Available Below
Product Details & Documents
HPLC LISA Waste bundle, type 20 with and without funnel.
Certifications: UN approved.
Delivery information: Kit includes 1×Safety waste cap LISA, S60/61, 1×20 L canister, S60/61, PE-HD, 1×exhaust filter M, with splash guard and change indicator.
Product Family Options
Product Information
- MaterialCapacityAvailabilityPrice
- 77553-672HPLC LISA Waste bundle with 1×funnel extension LISA, V3.0, type 20, Thread size: S60/61HPLC LISA Waste bundle with 1×funnel extension LISA, V3.0, type 20, Thread size: S60/61HDPE20 LLog in for availabilityEach$545.93Supplier Number: 75207
- 77553-670HPLC LISA Waste bundle, type 20, Thread size: S60/61Supplier Number: 75206