Waste Bundle for CPC Justrite Container
Catalog # 77553-676
Supplier: SCAT AMERICAS, INC.
New Product
Specifications
- Description:Waste bundle for justrite (CPC)
- Material:PTFE
- Cat. No.:77553-676
Specifications
About this item
Waste bundle for justrite (CPC).
Delivery: 1×HPLC manifold with CPC quick disconnect for justrite wast container, 1×Exhaust filter M, V3.0, with splash guard and change indicator, 1×Exhaust filter adapter to CPC coupling.