Cross Absorption Regeneration Buffer, 1X
Catalog # 77553-360
Supplier: Immunoreagents
New Product
CAS Number:
Specifications
- Description:Cross absorption regeneration buffer, proprietary buffer
- Size:500 ml
- Form:Liquid, 0.2 µm filtered
- Storage Temperature:2...8 °C
- Tested Applications:Cross absorption affinity purification
- Cat. No.:77553-360
Specifications
About this item
Caution: For in vitro laboratory use only. Not for diagnostic or therapeutic use. Not for human or animal consumption.