Orders LinkContactUsLinkComponent
Waste Bundle for Stainless Justrite Container
Waste Bundle for Stainless Justrite Container
Waste bundle for stainless justrite container, PTFE
Catalog # 77553-674
Supplier:   SCAT AMERICAS, INC.
Supplier #:  75208
Waste Bundle for Stainless Justrite Container
Catalog # 77553-674
Waste Bundle for Stainless Justrite Container
Waste bundle for stainless justrite container, PTFE
Price
$409.26
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Waste bundle for justrite (stainless).
Delivery information: Supplied with 1×HPLC manifold with stainless steel quick disconnect for justrite wast container, 1×Exhaust filter M, V3.0, with splash guard and change indicator, 1× Exhaust filter adapter to stainless steel coupling.
Specifications
  • Description:
    Waste bundle for stainless justrite container
  • Material:
    PTFE