SymLine HPLC Waste Bundles, V3.0
Supplier: SCAT AMERICAS, INC.
New Product
Certificates
About this item
SymLine FLEX bundle.
Certifications: UN-Y approved
Delivery: Supplied with 1×Basic 32 pipe, PE-HD electrostatic conductive, 1×GL 45 (m), 1×GL 25 (m), pipe connection (w), length: 310 mm, tensile strength (Rm) = 400 N, tightening torque = 4 Nm; 1×SymLine FlexTube, Length 1000 mm - flexible adaptable; 1×Shut-off, angled, 1×Exhaust filter M, V3.0; 1×Offset adapter 90° for exhaust filter; 1×Safety waste cap, S60/61, GL25 (m), connector for exhaust filter, PE-HD electrostatic conductive; 2×10 L / 2.64 gallon multilayer HDPE container with handle, natural colour. UN-Y approved, meets FDA standards; 1×Collecting tray with base insert; 1×Grounding cable; 1×LISA safety waste collector, GL45, collector for HPLC waste, 4×PFA-fittings for capillaries with (OD) 1,6/2,3/3,2 mm, 3×tube connector for tubes with (ID) 5,0 - 11,5 mm; 2×SymLine FLEX table mount.
Frequently Bought Together