Single Edge Blades, Unbacked, Unwrapped
Catalog # 77587-122
Supplier: Accutec Blades
New Product
Specifications
- Coating:Uncoated, oiled
- Material:Stainless steel
- Thickness:0.009"
- Cat. No.:77587-122
- Blade Edge:3 facet
- Backing material:Unbacked
Specifications
About this item
AccuTec Unbacked single edge blades feature 3 facet edges for sharpness.
- Unbacked blades
- Unwrapped blades for high volume use
- 3 facet edge for increased sharpness
- High quality stainless steel blade
- Made in USA
Packaging: Each box contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.