AccuTec Pro® Single Edge Blades, 3 Facet, Uncoated
Supplier: Accutec Blades
New Product
Certificates
About this item
AccuTec Pro® Single edge blades feature precision ground edges for sharpness and durability. Blades are wrapped in protective paper.
- 3 facet edge for increased sharpness
- Blades are individually wrapped for safety
- Made in USA
Reclosable clamshell for safe and convenient blade storage.
Packaging: Each clamshell contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Frequently Bought Together