Kinase Dilution Buffers VIII
About this item
Solution of buffer used for diluting kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Solution of buffer used for diluting kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Documents
TDS Kinase Dilution Buffer VIII
Datasheet
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77558-954
- 77558-956
- 77558-958
- Formulation:
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerol
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerol
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerol
- Storage Temperature:
- –20 °C
- –20 °C
- –20 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 77558-954Kinase dilution buffer VIIISupplier Number: K28-09-01Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-954FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerolStorage Temperature–20 °C - 77558-956Kinase dilution buffer VIIISupplier Number: K28-09-05Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-956FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerolStorage Temperature–20 °C - 77558-958Kinase dilution buffer VIIISupplier Number: K28-09-20Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-958FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerolStorage Temperature–20 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



