Kinase Dilution Buffers VI
About this item
Solution of buffer used for diluting kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Solution of buffer used for diluting kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Documents
TDS Kinase Dilution Buffer VI
Datasheet
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77558-942
- 77558-944
- 77558-946
- Formulation:
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerol
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerol
- Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerol
- Storage Temperature:
- –20 °C
- –20 °C
- –20 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 77558-942Kinase dilution buffer VISupplier Number: K26-09-01Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-942FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerolStorage Temperature–20 °C - 77558-944Kinase dilution buffer VISupplier Number: K26-09-05Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-944FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerolStorage Temperature–20 °C - 77558-946Kinase dilution buffer VISupplier Number: K26-09-20Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77558-946FormulationFrozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerolStorage Temperature–20 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



