Order Entry
Puerto Rico
ContactUsLinkComponent
 
Kinase Dilution Buffers V
New Product
 
Kinase Dilution Buffers V
New Product

 

  • Storage Temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 5 mM MgCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 5% glycerol