Kinase Dilution Buffers IV
New Product
Solution of buffer used for diluting kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Storage Temperature:–20 °C
- Formulation:Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 4 mM MgCl₂, 2.5 mM MnCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA