Bovine Galectin 1 (GAL1) ELISA Kit
Catalog # 77536-666
Supplier: Bioss
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Bovine
- Target protein:Galectin 1 (GAL1)
- Size:1 kit
- Detection Method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf Life:12 Months
- Detection Range:78.1 - 5000 pg/ml
- Storage Temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at −20 °C.
- Sensitivity:27.5 pg/ml
- Cat. No.:77536-666
- No. of tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results