New Product
Waste funnel bundle with 1x safety funnel 'LISA', V2.0.
Electrostatic conductive plastic: Yes
Grounding connection: Yes
Hinged lid: Yes
Main material: PE-HD-EL
Thread size: S60/61
: UN-Y approved.
Frequently Bought Together
Waste funnel bundle with 1x safety funnel 'LISA', V2.0.
Electrostatic conductive plastic: Yes
Grounding connection: Yes
Hinged lid: Yes
Main material: PE-HD-EL
Thread size: S60/61