Mini Industrial Vacuum Cleaner, VHS011 CIID2
Catalog # 77574-832
Supplier: Nilfisk
New Product
Specifications
- Vacuum Pressure:21 mbar
- Capacity:4 gal.
- Description:Mini industrial vacuum cleaner, VHS011 CIID2
- Environmentally Preferable:
- W×D×H:16.9×16.1×32.7"
- Frequency:60 Hz
- Noise level:72 dB(A)
- Power:0.95 kw
- Voltage:120 V
- Weight:46 lbs.
- Cat. No.:77574-832
Specifications
About this item
The VHS011 CIID2 is a range of CIID2-certified vacuum cleaners suitable for the collection of combustible dust and hazardous dust, in classified CIID2 areas.
- Lightweight and compact
- Highly movable due to the 4 ball bearing pivoting wheels
- Reliable with 950 W and great suction power
- Metal-motorhead integrates the brushless bypass motor both safer and more robust in massive usage than standard motors, moreover, it has an extended lifetime of up to 10.000 hours
- Fully equipped with convenient handle, power cord holder, and accessories holder
- Fully grounded and bounded, doesn't need inconvenient earthing wire and clamp
- Pull & Clean system grants high performance of filtration without usage of consumables
- 3 levels of safe disposal for the safest level of dust collection according to powder's classification, LCLASS
- IP65 water and dust resistance of the motor head grants the highest protection for operators and the environment
- Third-party certified CIID2 and LMH
This model VHS011, are perfect for cleaning jobs in the food industry, where most of the powders are combustible, designed to be used for cleaning food processing machines e.g. vertical packaging machines, conveyors belts, V-blenders, etc. and for general cleaning of floors, desks, equipment etc.